Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Brand:  Novus Biologicals™ NBP2-31142SS
Product Code. 18113923
  • 172.00 € / 25µg
Explore more special offers
Specifications
1.0 mg/mL
0.025 mg
CpG Oligodeoxynucleotides Mouse
Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 μg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.)
CpG Oligodeoxynucleotides