Learn More
Control Company™ Cradle and USB Cable for Logger-Trac™ Temp Mini Datalogger
Use for downloading temperature reading from Logger-Trac™ Temp Mini Datalogger
Brand:  Control Company™ 61012-10
Product Code. 15586429
  • 45.86 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Required accessory for the Logger-Trac™ Mini Datalogger.
  • Cradle is constructed of plastic
  • Includes USB cable for downloading data onto datalogger
Specifications
Specifications
USB cable
Plastic
Logger-Trac™ Temp Mini Datalogger