missing translation for 'onlineSavingsMsg'
Learn More

Creatine monohydrate, 99%

Product Code. 10783421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10783421

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Creatine monohydrate, CAS # 6020-87-7, is the monohydrate form of creatine, an amino acid found primarily in skeletal muscle cells that is important for rapid energy replenishment. | CAS: 6020-87-7 | C4H9N3O2·H2O | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Creatine is an amino acid made naturally by the body that can also be acquired through dietary consumption
  • It is primarily stored in skeletal muscle tissue
  • Creatine is stored in a phosphate form, and upon depletion of ATP deposits, phosphocreatine is able to donate phosphate groups to rapidly replenish ATP
  • Creatine supplementation may be beneficial as a disease treatment, particularly in diseases of muscle wasting including cancer and chronic disease

Applications

  • Creatine may be used in the laboratory to study its potential for use as a treatment for metabolic and muscle wasting diseases
  • Creatine has been found to have antitumor, antiviral, and antidiabetic effects
  • It is also used in laboratory settings to study neurological disorders and has been found to be neuroprotective against hypoxic conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6020-87-7
Molecular Formula C4H9N3O2·H2O
Molecular Weight (g/mol) 149.15
MDL Number MFCD00071582
InChI Key MEJYXFHCRXAUIL-UHFFFAOYSA-N
Synonym creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure
PubChem CID 80116
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=N)N.O

Specifications

Melting Point 292.0°C
Color White to Yellow
Ignition Residue 1% max.
Loss on Drying 12.6% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (HPLC)
Packaging Glass bottle
Linear Formula H2NC(=NH)N(CH3)CH2CO2H·H2O
Quantity 100 g
Beilstein 04, 363
Merck Index 14, 2568
Solubility Information (1% in water) Clear colorless to light yellow
Formula Weight 149.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Creatine monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.