missing translation for 'onlineSavingsMsg'
Learn More
Creatinine, 98%, Thermo Scientific Chemicals Product Code.: 11916261

Creatinine, 98%, Thermo Scientific Chemicals

Product Code. 11916261
250 g, Glass bottle
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11916261

Brand: Thermo Scientific Acros 228942500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point 255.0°C
Color White
Flash Point 290°C
Loss on Drying 0.5% (105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 g
Beilstein 24,245
Merck Index 15,2557
Solubility Information (5% in water) Clear colorless to slightly yellow
Formula Weight 113.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Creatinine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469079 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number228940500,228942500,228940250
Lot NumberA0458263 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number228940500,228942500,228940250
Lot NumberA0457392 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number228940500,228942500,228940250
Lot NumberA0455987 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number228940500,228942500,228940250
Lot NumberA0453668 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number228940500,228942500,228940250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Creatinine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.