missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, Indicator Grade, Pure

30.55 € - 234.17 €

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10467740
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
151380050
5 g Glass bottle
30.55 €
5g
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
10103350
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
151380250
25 g Glass bottle
84.52 €
25g
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
10704294
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
151381000
100 g Glass bottle
234.17 €
100g
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1733-12-6
382.43
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CHEBI:86218
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
C21H18O5S
MFCD00005878
73013
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol

Specifications

290.0°C
C21H18O5S
19, 91
Solubility in water: soluble. Other solubilities: soluble in dil. acids, dil. alkalies and ethanol,, insoluble in diethyl ether, benzene
Authentic
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
73013
382.42
1500 min. (572nm, in 0.01N NaOH)
Brown/Red or Green
Cresol Red
1733-12-6
MFCD00005878
15, 2568
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
382.43
CHEBI:86218
Pure
from pH 7.0 (orange) to pH 8.8 (purple)
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cresol Red

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.