missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Crimping Heads for Programmable Electronic High Power Crimp Station

For use with Programmable Electronic High Power Crimp Station

414.12 € - 443.00 €

Specifications

Material Plastic, Steel
View More Specs

Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15532340
View Documents
Fisherbrand™
20 06 3206
20mm Crimping Head This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15542300
View Documents
Fisherbrand™
08 06 3200
Crimping Head for 8mm Aluminum Caps
443.00 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
15542310
View Documents
Fisherbrand™
11 06 3202
11mm Crimping Head This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15562330
View Documents
Fisherbrand™
13 06 3204
13mm Crimping Head This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Crimping Head for Aluminum Caps
Specifications

Specifications

Plastic, Steel
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.