missing translation for 'onlineSavingsMsg'
Learn More

Croconic acid, 97%, Thermo Scientific™

Product Code. 10401533
Click to view available options
Glass bottle; 100mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 488-86-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 331.0°C to 334.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H2O5
MDL Number MFCD00181389
Quantity 100 mg
Synonym croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
Solubility Information Solubility in water: scarcly soluble. Other solubilities: soluble in methanol
InChI Key RBSLJAJQOVYTRQ-UHFFFAOYSA-N
SMILES C1(=C(C(=O)C(=O)C1=O)O)O
IUPAC Name 4,5-dihydroxycyclopent-4-ene-1,2,3-trione
Molecular Weight (g/mol) 142.07
PubChem CID 546874
Formula Weight 142.07
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Croconic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'