missing translation for 'onlineSavingsMsg'
Learn More
Croconic acid disodium salt, 97%, Thermo Scientific Chemicals Product Code.: 11475876

Croconic acid disodium salt, 97%, Thermo Scientific Chemicals

Product Code. 11475876
1 g
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy

Product Code. 11475876

Brand: Thermo Scientific Alfa Aesar B22235.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14379-00-1
Molecular Formula C5Na2O5
Molecular Weight (g/mol) 186.03
MDL Number MFCD00191954
InChI Key OQXLFPHHAAAVKQ-UHFFFAOYSA-L
Synonym croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt
PubChem CID 12120285
IUPAC Name disodium;3,4,5-trioxocyclopentene-1,2-diolate
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 186.03
Percent Purity 97%
Chemical Name or Material Croconic acid disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Croconic acid disodium salt, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.