Description
- Guaranteed inert and leak-free
- Individually polished to stay cleaner for longer
Order Info
Includes
Specifications
Autoclavable | |
White | |
195 x 95mm | |
30mm | |
95mm | |
1/Ea. |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
Containers & Vessels / Stirrer Hot Plates | |
5 Cross Stirrer Bars 1 each of 10x10, 20x20, 25x25, 30x30, 38x38 mm | |
195 mm |