missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Crucible
Bochem™ Crucible is constructed of Nickel 99.5%.
Brand: Bochem™ 8103
This item is not returnable.
View return policy
Description
- Nickel 99.6%
- Lids to be ordered separately
Specifications
Nickel | |
25 mL | |
35mm | |
35mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction