missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ CRUCIBLE TONGS PTFE Coated TIPS 18in

Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.

Brand:  Fisherbrand™ 13-820-022

Product Code. 17801588

  • 270.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.

  • Stainless steel construction
  • PTFE coating
  • Nonsticking and non-corrosive

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Specifications

Crucible tongs
18 in.
PTFE
457 mm
Crucible Tongs
Tongs with PTFE coating
1
Ribbed
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.