Learn More
Tempshield™ Cryo-Gloves™ Blue Mid-Arm Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
320.00 € - 337.00 €
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Mid-Arm Length |
Color | Blue |
Hand | Specific |
Product Type | Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11803801
|
Tempshield™
MAM |
Medium |
320.00 €
1 pair Qty Price Break Available |
In Stock
Log in to see stock. ![]() |
||||||
11813801
|
Tempshield™
MAL |
Large |
320.00 €
1 pair Qty Price Break Available |
In Stock
Log in to see stock. ![]() |
||||||
16002232
|
Tempshield™
MAXL |
X-Large |
337.00 €
1 pair Qty Price Break Available |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Cryogenic Gloves | |
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems | |
-320°F | |
13.3 to 15.5 in. |
Mid-Arm Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
33.5 to 39.5 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.