missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Waterproof Mid-Arm Length Cryogenic Gloves

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

Brand:  Tempshield™ MAXLWP

335.79 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15210126

  • 384.00 € / 1 pair
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

This item is not returnable. View return policy

Descrizione

Descrizione

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • Thin, seamless 100% waterproof liner provides protection from spills and splashes
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifica

Specifica

EN 420, EN 511
Blue
Waterproof Cryogenic Gloves
Yes
Cryo-Gloves™
-196°C
33.5 to 39.5 cm
Mid-Arm Length
Specific
X-Large
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes
-320°F
13.3 to 15.3 in.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali