missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Gloves - Elbow Length

Product Code. 15499509
Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY

Specifications

Description Elbow Length
Color Blue
Hand Specific
Size Large
For Use With (Application) Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English) 18 in.
Length (Metric) 45.7 cm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'