missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Gloves - Shoulder Length

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

Specifications

Description Shoulder Length
Color Blue
For Use With (Application) Handling Samples from Ultra-Low Temperature and Cryogenic Storage
Length (English) 28 in.
Length (Metric) 71.12 cm
View More Specs

Description

Description

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
Specifications

Specifications

Shoulder Length
Handling Samples from Ultra-Low Temperature and Cryogenic Storage
71.12 cm
Blue
28 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.