missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Protection™ Face Shield

Product Code. 15801787
Click to view available options
:
Face Shield; Color: Clear
This item is not returnable. View return policy

Product Code. 15801787

Brand: Tempshield™ FSH1001

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Molded polycarbonate Cryo-Protection™ Face Shield is shaped to hug your chest and face, and pivots up and out of the way when not in use. The padded head gear has a ratchet band and pin-and-lock system for a 2-way adjustable fit.

  • Injection molded polycarbonate shield
  • Hugs your chest and the sides of your face
  • Pivots up, away from your face when not in use
  • Ratchet band and pin-and-lock systems provides a 2-way
  • Adjustable fit
  • Lightweight design
  • CE Cat. II
  • Impact resistant after being splashed with liquid cryogens
  • Retains optical clarity
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Lens Type Splash Protection
Lens Coating Impact-resistant
Color Clear
Certifications/Compliance (Europe) CE Category II, Meets ANSI Z87.1
For Use With (Application) Handling Cryogenic Liquids
Frame Color Black
Lens Material Polycarbonate
Frame Type Padded Head Gear with Ratchet Band
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.