Learn More
Thermo Scientific™ CryoBox™ Freezer Racks
GREENER_CHOICE
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Brand:  Thermo Scientific™ 5038-4422
401.17 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 10653791
  • 521.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
  • Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
  • Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
  • Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Silver
16 CryoBoxes/Rack
Horizontal
Cryogenic Box Storage Rack, Horizontal
5.625 x 8.875 x 23.25 in. (22.5 x 59.0 x 14.3 cm)
Each
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ CryoBox™ Freezer Racks > Horizontal, stainless-steel, 16 compartments, 5.625 x 8.875 x 23.25 in. (D x W x H)