Learn More
R&D Systems™ CryoDefend-Cell Lines (5 x 10 mL)
Media for defined, protein-free cryopreservation of cultured cells
Brand: R&D Systems™ CCM019
172.78 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Includes: 5 x 10mL vials CryoDefend-Stem Cells Media
Description
Cryopreservation can be used to maintain cells at a low passage number and to reduce the risk of contamination by microbes and other cell types. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
Contains a defined serum substitute as well as an optimized concentration of a cryopreservative that increases the recovery and viability of healthy cells compared to conventional freezing media
This product contains
DMSO and is ready for use
Reduces experimental variability by using fully defined media
Suitable for all cell types
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
1 Kit |