Learn More
R&D Systems™ CryoDefend-Stem Cells (5 x 10 mL)
Media for defined, protein-free cryopreservation of stem cells
Brand:  R&D Systems™ CCM018
228.07 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18491540
  • 290.00 € / 1 set
Explore more special offers
Description
Description
Cryopreservation is essential for the banking of valuable stem cell cultures. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
  • Reduce experimental variability by using fully defined media
  • Supports post-thaw cell viability better than conventional freezing media
  • Optimized for cryopreservation of stem cells
  • Suitable for all stem cell types
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
CryoDefend-Stem Cells
5 x 10 mL