Learn More
SPEX Certiprep™ Cryogenic Gloves
Product Code. 13463530
Click to view available options
Size:
Small
Medium
Large
SPEX Certiprep™ Cryogenic Gloves are designed to protect hands and arms from ultra-cold materials and liquid nitrogen exposure.
  • Cryogenic gloves specifically designed to protect hands and arms from the hazards associated with liquid nitrogen handling
  • Highly recommended for the safe operation of the Freezer/Mill™
  • Multi-layer insulation for optimal protection
  • Flexible and lightweight to allow excellent dexterity
  • Comfortable to wear for extended periods of time
Specifications
Color Blue
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gloves
Size Medium
Sterility Non-sterile