Learn More
Fisherbrand™ Cryogenic Labels for Cryogenic Storage
GREENER_CHOICE
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy-to-use dispenser box with 20 sheets per pack.
Brand:  Fisherbrand™ 15920A
12.93 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17867281
  • 16.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to +121°C (-321° to +250°F)
  • No need to self-laminate
  • Thinner labels allow tubes to fit most storage systems
  • Available in a full range of colors and sizes
  • Guaranteed not to fall off under normal use (provided user instructions are followed)
  • Also offered in laser printer-compatible sheets (see Cat. Nos. 15-930 and 15-940 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifications
Specifications
White
Cryogenic Label
0.5 to 1.5mL tube
0.375 in. dia.
-196°C to +121°C
0.375 in.
Label
Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
1000/Roll
-321 to 250°F
0.375 in. dia.
0.95 cm