Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
Perform controlled-rate freezing applications with a controlled-rate freezer and these CryoMed bag freezing presses.
Brand:  Thermo Scientific™ 4000555
 View more versions of this product
Product Code. 11746499
  • 904.50 € / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform controlled-rate freezing applications with a controlled-rate freezer and these Thermo Scientific™ CryoMed™ Bag Freezing Presses.
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2 to 4 bags.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hält 2 Beutel
Nexell R4R9955 (Fenwal 4R5462)
CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Bags (2)