Learn More
Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
127.64 € - 537.68 €
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10795505
View Documents
Thermo Scientific™
4000710
Straw Holder
537.68 €
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
11859480
View Documents
Thermo Scientific™
4000711
Vial Holder
506.52 €
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
11869480
View Documents
Thermo Scientific™
4000714
Cane holder for 1.2 and 2.0 vials
127.64 €
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
Description
Description
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
  • Warranty varies by country, please contact us for details
  • Videos