Learn More
Thermo Scientific™ CryoMed™ with OPC UA, General Purpose

Thermo Scientific CryoMed Controlled-Rate Freezers with OPC UA are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
39496.50 € - 44622.00 €
Specifications
Temperature Range | -180°C to +50°C |
---|---|
Cabinet Material | Stainless Steel |
Sample Loading | Front-loading |
Data Outputs | USB and OPC UA (ethernet) |
On-Board Datalogging | Yes |
Product Code | Brand | Capacity (Metric) | Type | Electrical Requirements | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Type | Electrical Requirements | Price | Quantity & Availability | ||||
17445173
![]() |
Thermo Scientific™
TSCM34FV |
34 L | Controlled-rate Freezer with Printer | 220-230V, 50-60Hz |
42712.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17535865
![]() |
Thermo Scientific™
TSCM34SV |
34 L | Controlled-rate Freezer with Printer | 220-230 V, 50-60 Hz |
41506.50 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17545865
![]() |
Thermo Scientific™
TSCM48FV |
48 L | Controlled-rate Freezer with Printer | 220-230V, 50-60Hz |
44622.00 €
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||
17555865
![]() |
Thermo Scientific™
TSCM48SV |
48 L | Controlled-rate Freezer without Printer | 220-230V, 50-60Hz |
43315.50 €
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
17515865
![]() |
Thermo Scientific™
TSCM17FV |
17 L | Controlled-rate Freezer with Printer | 220-230V, 50-60Hz |
40702.50 €
Each |
Estimated Shipment: 08-09-2025
Log in to see stock. |
||||
17525865
![]() |
Thermo Scientific™
TSCM17SV |
17 L | Controlled-rate Freezer without Printer | 220-230V, 50-60Hz |
39496.50 €
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
Description
For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
User-friendly operation with enhanced data traceability
- Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
- OPC UA (ethernet) serial communication protocol
- Factory-issued certificates
- Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
- Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
- USB data export of .pdf file run logs support 21 CFR part 11
- User security: 3 levels of user accessibility
- “Run last” feature allows the same profile to be run consecutively with the touch of a button
- PC interface software provides remote operation capabilities, run review, and custom profile creation
Real-time run monitoring for sample protection
- Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
- Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
- Remote alarm contacts
- Optional thermal printer
Reliable temperature performance for high throughput usage
- Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
- Preventative maintenance indicator for LN2 solenoid replacement limits downtime
- Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
- Compatible with VHP cleaning techniques
- Environmentally friendly insulating foam
- Type 304 stainless steel with exterior powder-coat finish
Warranty varies by region. Check with your local representative for more information.
Services and Support (optional)
When you invest in Thermo Scientific laboratory equipment, you not only benefit from top-quality product performance, but you can also enjoy peace of mind and maximum uptime with world-class service solutions provided by Unity Lab Services.
Service plans* – Maximize uptime, maintain peak performance, and extend the life of your equipment with a service plan that includes annual preventive maintenance, digital remote resolution, and fast onsite response times.
Catalog numbers:
- Factory Warranty Period
- Total Care Warranty TCWTYSCRYO
- Post Warranty Period
- Extended Warranty EXTWSCRYO
- Tech Direct Service Plan TEDSCRYO
- Total Care Service Plan TLCSCRYO
Compliance Services** – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Catalog numbers:
Qualification service
- On-site IQ/OQ service IOQP003507
- On-site IQ/OQ + temperature mapping IOTQP003507
- On-site temperature mapping service TEMPMAP
*Available in 23 countries. Please indicate your interest in the “Request a quote” form and we will come back to you on availability.
**Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Specifications
-180°C to +50°C | |
Front-loading | |
Yes | |
Thermo Fisher Connect compatible |
Stainless Steel | |
USB and OPC UA (ethernet) | |
PC Interface software supported |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.