missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryoscope Standard 530 (-0.512°C) (530m°H), Reagecon
Used to calibrate, control, verify, or validate cryoscopes.
Marke: Reagecon MTR03025
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
- Cryoscope Reference Standard, Cryoscope Reference Material
- Used to calibrate, control, verify or validate cryoscopes.
- Manufactured for use with all cryoscopes compliant to International Reference Standard ISO5764/IDF108 for the determination of freezing point in milk.
- Prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
- Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The certified values of each standard are verified using a high performance Cryoscope calibrated with NIST traceable, ISO Guide 34 Certified Standards.
Specifications
HDPE Flip-Top Bottle | |
530 (−0.512°C) (530m°H) | |
Cryoscope | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction