missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptotanshinone, MedChemExpress
MedChemExpress Cryptotanshinone is a natural compound extracted from the root of Salvia miltiorrhiza Bunge that shows antitumor activities. Cryptotanshinone inhibits STAT3 with an IC50 of 4.6 μM.
72.90 € - 261.90 €
Specifications
Chemical Name or Material | Cryptotanshinone |
---|---|
CAS | 35825-57-1 |
Color | Orange |
Purity Grade Notes | Research |
Molecular Formula | C19H20O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16438888
|
Medchem Express
HY-N0174 |
10 mg |
72.90 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16464777
|
Medchem Express
HY-N0174 |
50 mg |
261.90 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cryptotanshinone is a natural compound extracted from the root of Salvia miltiorrhiza Bunge that shows antitumor activities. Cryptotanshinone inhibits STAT3 with an IC50 of 4.6 μM.Specifications
Cryptotanshinone | |
Orange | |
C19H20O3 | |
DMSO : 2 mg/mL (6.75 mM; ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble) | |
296.36 | |
98.02% | |
Powder |
35825-57-1 | |
Research | |
Cryptotanshinon Tanshinone c | |
O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC[C@@H]4C)=O | |
296.36 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
H301∣H410
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only