missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal violet, ACS reagent

58.89 € - 190.95 €

Specifications

CAS 548-62-9, 90-94-8
Melting Point 173°C
Molecular Formula C25H30ClN3
Merck Index 15,443
Synonym crystal violet, gentian violet, basic violet 3, methylrosaniline chloride, aniline violet, hexamethyl violet, gentiaverm, hexamethylpararosaniline chloride, pyoktanin, adergon
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11976931
View Documents
Thermo Scientific Chemicals
447570500
50 g Glass Bottle
58.89 €
50g
Please to purchase this item. Need a web account? Register with us today!
11986931
View Documents
Thermo Scientific Chemicals
447572500
250 g Glass bottle
190.95 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

548-62-9, 90-94-8
C25H30ClN3
crystal violet, gentian violet, basic violet 3, methylrosaniline chloride, aniline violet, hexamethyl violet, gentiaverm, hexamethylpararosaniline chloride, pyoktanin, adergon
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
173°C
15,443
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform,1g/10 mL alcohol,1g/15 mL glycerin,practically insoluble in ether
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.99
CHEBI:41688
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-953-6

Product Identifier
  • Crystal Violet

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : C.I. Basic Violet 3

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.