missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Crystal Violet, certified
Brand: Thermo Scientific Chemicals 405835000
233.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
Crystal Violet | |
548-62-9, 90-94-8 | |
88% min. | |
MFCD00011750 | |
15, 4430 | |
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether | |
Authentic | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.99 | |
Glass bottle | |
500 g |
215.0°C | |
0.98 to 1.2 (P-15/P+15) | |
C25H30ClN3 | |
13, 756 | |
Basic Violet 3, C.I. 42555, Gentian Violet | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
Certified | |
Green | |
Crystalline Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction