Learn More
Thermo Scientific Chemicals Crystal Violet, pure, indicator
Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer.
31.60 € - 266.00 €
Specifications
Chemical Name or Material | Crystal Violet |
---|---|
Melting Point | 215.0°C |
Name Note | Indicator, Pure |
CAS | 548-62-9, 90-94-8 |
Molecular Formula | C25H30ClN3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10646972
|
Thermo Scientific Chemicals
212120250 |
25 g | Glass bottle |
31.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637884
|
Thermo Scientific Chemicals
212121000 |
100 g | Glass bottle |
78.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177663
|
Thermo Scientific Chemicals
212125000 |
500 g | Glass bottle |
266.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
Crystal Violet | |
Indicator, Pure | |
C25H30ClN3 | |
13, 756 | |
Basic Violet 3, C.I. 42555, Gentian Violet | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.99 | |
Glass bottle |
215.0°C | |
548-62-9, 90-94-8 | |
MFCD00011750 | |
15, 4430 | |
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
Pure |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Crystal Violet
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : C.I. Basic violet 1,Michler's ketone
RUO – Research Use Only