missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSF2RA/CD116 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18307-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.Specifications
CSF2RA/CD116 | |
Polyclonal | |
Unconjugated | |
CSF2RA | |
CD116, CDw116, CSF2R, CSF2RA, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM CSF R alpha, GMCSFR, GMCSFR alpha, GMR, GMR alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1438 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15509 | |
CSF2RA | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |