missing translation for 'onlineSavingsMsg'
Learn More

CSF2RA/CD116 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18307-1-AP-20UL

 View more versions of this product

Product Code. 16858384

  • 145.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.
Specifications

Specifications

CSF2RA/CD116
Polyclonal
Unconjugated
CSF2RA
CD116, CDw116, CSF2R, CSF2RA, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM CSF R alpha, GMCSFR, GMCSFR alpha, GMR, GMR alpha
Rabbit
Antigen Affinity Chromatography
RUO
1438
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15509
CSF2RA
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers