missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CSNK1G2 (CK1 gamma 2), His Tag Recombinant Protein
Human CSNK1G2 (CK1 gamma 2), His Tag Recombinant Protein
Brand: Thermo Scientific™ PV3499
This item is not returnable.
View return policy
Description
- A serine/threonine kinase implicated in a wide range of cellular functions including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis
Specifications
NP_001310 | |
1455 | |
10 μg | |
CK1 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.04% Triton X-100, 4 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser/Thr Kinases: CK1 Group | |
Zin-LYTE™, Omnia™ Bulk Substrate | |
casein kinase 1, gamma 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
52.5 kDa | |
CSNK1G2 (CK1 γ 2) | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
CSNK1G2 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |