missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK2A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10606-1-AP-150UL
This item is not returnable.
View return policy
Description
Casein kinase II (CK2) is a constitutively active, ubiquitously expressed serine/threonine protein kinase that is thought to have a regulatory function in cell proliferation, cell differentiation and apoptosis. CK2 functions as a tetrameric complex consisting of two regulatory beta-subunits and two catalytic units (alpha and alpha′) in a homomeric or heteromeric conformation. Whilst the alpha- and alpha′-subunits are catalytically identical, proteins that regulate CK2, such as cdc2 and Hsp90, preferentially bind to the alpha and not the alpha′-subunit. CK2 can phosphorylate a number of key intracellular signaling proteins implicated in tumor suppression (p53 and PTEN) and tumorigenesis (myc, jun, NF-kappaB). CK2 is also thought to influence Wnt signaling via beta-catenin phosphorylation and the PI 3-K signaling pathway via the phosphorylation of Akt.Specifications
CSNK2A2 | |
Polyclonal | |
Unconjugated | |
CSNK2A2 | |
Casein kinase II subunit alpha, CK II alpha, CK2A2, CSNK2A1, CSNK2A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1459 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19784 | |
CSNK2A2 | |
CSNK2A2 Fusion Protein Ag0877 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |