missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Immune Cell Serum Replacement (SR)

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells when added as a serum substitute to supplement a basal cell culture medium such as Gibco CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium.

Brand:  Gibco™ A4702902

 View more versions of this product

Product Code. 17777187

  • 3655.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store flat at -5°C to -20°C.
Xeno-free
Sterile-filtered
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Dry Ice
Human Immune Cells
Serum Replacement
Bag - BioProcess Container (BPC)
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2477504 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA4702901,A4702902
Lot Number2631957 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA4702901,A4702902
Lot Number2610209 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA4702901,A4702902
Lot Number2805612 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA4702901,A4702902
Lot Number2824440 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA4702901,A4702902
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CTS™ Immune Cell Serum Replacement (SR) > Quantity: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.