missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ PSC Cryopreservation Kit

Product Code. 17134719
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Artikelnummer. 17134719

Brand: Gibco™ A4239301

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

The CTS PSC Cryopreservation Kit is cGMP manufactured and used for the cryopreservation and recovery of pluripotent stem cells (PSCs).

The CTS PSC Cryopreservation Kit is cGMP manufactured and used for the cryopreservation and recovery of pluripotent stem cells (PSCs). The kit consists of CTS PSC Cryomedium, a xeno-free solution for the cryopreservation of early passage PSCs, and CTS RevitaCell Supplement (100X), a chemically defined recovery supplement for use in the post-thaw culture medium. In combination, these reagents help minimize loss of cell viability, maximize post-thaw recovery, and minimize unwanted differentiation of pluripotent stem cells.

The CTS PSC Cryopreservation Kit:
• Provides efficient recovery and higher viability than traditional methods of PSC cryopreservation
• Facilitates regulatory filings through cGMP manufacturing and the availability of documentation
• Simplifies the transition from bench to clinic through the direct replacement of the RUO PSC Cryopreservation Kit

Efficient recovery
The CTS PSC Cryopreservation Kit achieves higher viability than traditional methods of cryopreservation. It is compatible with clump and single cell passaged PSCs cultured in a variety of media systems. CTS PSC Cryomedium is provided as a ready-to-use, complete solution containing 10% DMSO for efficient cell addition. Separate additions of enhancing reagents are not required, helping save valuable preparation time.

Facilitates regulatory filings
The CTS PSC Cryopreservation Kit is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability and convenient access to our Drug Master File (DMF) are available for the CTS PSC Cryopreservation Kit.

Simplifies transition from bench to clinic
The CTS PSC Cryopreservation Kit provides the same efficient recovery, consistency, and reduced variability as the RUO PSC Cryopreservation Kit. With its additional safety testing, including adventitious agents, the CTS PSC Cryopreservation Kit enables a smooth transition from the RUO PSC Cryopreservation Kit.

TRUSTED_SUSTAINABILITY

Tekniske data

Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Xeno-free
Form Liquid
Product Type Cryopreservation Medium
Sterility Sterile-filtered
With Additives DMSO (10%)
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 1 Kit
Shipping Condition CTS PSC Cryomedium: Dry Ice
CTS RevitaCell: Dry Ice
Format Bottle
Content And Storage • 50 mL CTS PSC Cryomedium; store at –5 to –20°C and protect from light
• 5 mL CTS RevitaCell Supplement (100X); store at –5 to –20°C and protect from light
Vis mere Vis mindre

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.