missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CU-CPT17e, MedChemExpress
MedChemExpress CU-CPT17e is a potent multi-Toll-like receptor (TLR) agonist that activates TLR3, TLR8, and TLR9.
716.85 € - 4249.80 €
Specifications
Chemical Name or Material | CU-CPT17e |
---|---|
CAS | 2109805-75-4 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C27H24N2O8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16492806
|
Medchem Express
HY-101929 |
25 mg |
1926.45 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16484726
|
Medchem Express
HY-101929 |
100 mg |
4249.80 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460037
|
Medchem Express
HY-101929 |
50 mg |
3121.20 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16446377
|
Medchem Express
HY-101929 |
5 mg |
716.85 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16423648
|
Medchem Express
HY-101929 |
10 mg |
1062.45 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CU-CPT17e is a potent multi-Toll-like receptor (TLR) agonist that activates TLR3, TLR8, and TLR9.Specifications
CU-CPT17e | |
Light Yellow | |
C27H24N2O8 | |
O=[N+]([O-])C(C=C1)=CC=C1COC2=CC(C=CC3(CCOCC3)O4)=C4C=C2OCC5=CC=C([N+]([O-])=O)C=C5 | |
504.49 | |
Research | |
Cancer-programmed cell death |
2109805-75-4 | |
Research | |
DMSO : 5 mg/mL (9.91 mM; Need ultrasonic and warming) | |
504.49 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only