Learn More
Sartorius Cubis™ II High-Capacity Micro Balance, 36S
GREENER_CHOICE
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Brand:  Sartorius MCA36S-3CEU-D QP99
 View more versions of this product
Product Code. 17907400
  • 37160.00 € / Each
Explore more special offers
Description
Description
  • Ultra-high resolution with up to 61 million weighing steps (digits).
  • End-to-end data integrity.
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management.
  • Cleanability, part of compliance.
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling.
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer.
  • Optional inner draft shield for best weighing performance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MCA
Color Touchscreen
36S Weighing Module, All-Inclusive Package QP99
3.5 sec.
9.4 in.
11.9 in.
32 g
0.001 mg
MCA36S-3CEU-D QP99
Balance
240 mm
302 mm