Learn More
Sartorius Cubis™ II Micro Balance, MCA User Interface
GREENER_CHOICE
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCA10.6S-M-MTS
 View more versions of this product
Product Code. 30052880
  • 46485.00 € / Each
Explore more special offers
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor
    (draft shield M), or gesture sensor (optional) learning capability
Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
21 CFR PART 11 17025 USP 41
10.1 g
1 μg
±3 μg
Draft shield M: Motorized
Micro Balance
340/355 mm weighinh/electrical module
129/61 mm weighing/electrical module
100/240 V (±10%)
1 USB B port, 1 USB C Port, 1-9 pin RS 232 connection, 1 ethernet port
Color TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±5 μg
10 years
139 mm for weighing/240 mm for electrical module
7.2, 7.9 kg