missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis II MTS Balance
GREENER_CHOICE

The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.

Brand:  Sartorius MCA8202S-0-MTS

 View more versions of this product

Product Code. 30052870

  • 11380.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Leading performance.
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling
  • The modular weighing system Cubis™ II can be individually configured from different components.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

D-sub connector, 9-pin
8200 g
10 mg
Cubis II MTS Balance
Balance
Class II
MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
Color Touchscreen
Die-cast Aluminum, Plastic, Optiwhite Float Glass and Stainless Steel
1 second
00 to 240 VAC (±10%)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers