Learn More
Sartorius Cubis II MTS Balance
GREENER_CHOICE
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.
7930.00 € - 15375.00 €
Especificaciones
Connection Type D-sub connector, 9-pin
Display Type Color Touchscreen
Model Cubis II MTS Balance
Type Balance
Electrical Requirements 00 to 240 VAC (±10%)
Ver más especificaciones
Productos 3
Código de producto Marca Capacity Readability Description Precio Cantidad y disponibilidad  
Código de producto Marca Capacity Readability Description Precio Cantidad y disponibilidad  
30052872
GREENER_CHOICE
missing translation for 'viewDocuments'
Sartorius
MCE8202S-0-MTS
8200 g 10 mg MCE Display with factory-installed essential weighing applications
7930.00 €
Each
30052870
GREENER_CHOICE
missing translation for 'viewDocuments'
Sartorius
MCA8202S-0-MTS
8200 g 10 mg MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
11380.00 €
Each
30052885
GREENER_CHOICE
missing translation for 'viewDocuments'
Sartorius
MCA125S-U-MTS
120 g 0.01 MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
15375.00 €
Each
Descripción
Descripción
  • Leading performance.
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling
  • The modular weighing system Cubis™ II can be individually configured from different components.
Especificaciones
Especificaciones
D-sub connector, 9-pin
Cubis II MTS Balance
00 to 240 VAC (±10%)
Color Touchscreen
Balance
Videos
SDS
Documentos
Documentos
Certificados