Learn More
Sartorius Cubis™ II Precision (2-place) Balance, MCE User Interface
BIOPHARMAGREENER_CHOICE
Cubis™ II Precision (2-Place) Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCE8202S-2S00-0
 View more versions of this product
Product Code. 16407495
  • 7475.00 € / Each
Explore more special offers
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±4 mg
Precision (2-place) Balance
425 mm
95 mm
120/240 V
50/60 Hz (± 5%)
8200 g
0.01 g
±6 mg
No draft shield
1 sec
10 years
240 mm
5.4 kg
100/240 V