Learn More
Sartorius Cubis™ II Precision Lab Balance
Ideal model for every application with a maximum load between 320 g and 14,200 g and a readability of 1 mg to 100 mg.
7470.00 € - 15545.00 €
Specifications
Certifications/Compliance | Balance with Type Approval Certificate for China |
---|---|
Connection Type | D-sub connector, 9-pin |
Display Type | Color Touchscreen |
Includes | Internal Calibration |
Material | Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel |
Product Code | Brand | Capacity | Readability | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Model | Price | Quantity & Availability | |||
16303297
|
Sartorius
MCA6202P-2S00-0 QP99 |
1500 g, 3000 g, 6200 g | 10 mg, 20 mg, 50 mg | Balance without Draft Shield |
8455.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16352276
|
Sartorius
MCA14202S-2S00-0 QP99 |
14,200 g | 10 mg | Balance without Draft Shield |
15545.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16413275
|
Sartorius
MCA14202P-2S00-0 QP99 |
3500 g, 7000 g, 14,200 g | 10 mg, 20 mg, 50 mg | Balance without Draft Shield |
12900.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16415435
|
Sartorius
MCA5201S-2S00-0 QP99 |
5200 g | 100 mg | Balance without Draft Shield |
7470.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16415615
|
Sartorius
MCA5202S-2S00-E QP99 |
5200 g | 10 mg | Balance with Draft Shield E |
10365.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16415645
|
Sartorius
MCA5202S-2S00-R QP99 |
5200 g | 10 mg | Balance with Draft Shield R |
10365.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16416735
|
Sartorius
MCA623P-2S00-E QP99 |
150 g, 300 g, 620 g | 1 mg, 2 mg, 5 mg | Balance with Draft Shield E |
9010.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16416765
|
Sartorius
MCA623P-2S00-R QP99 |
150 g, 300 g, 620 g | 1 mg, 2 mg, 5 mg | Balance with Draft Shield R |
9010.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16424855
|
Sartorius
MCA323S-2S00-E QP99 |
320 g | 1 mg | Balance with Draft Shield E |
9260.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16424885
|
Sartorius
MCA323S-2S00-R QP99 |
320 g | 1 mg | Balance with Draft Shield R |
9260.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16442465
|
Sartorius
MCA10202S-2S00-0 QP99 |
10,200 g | 10 mg | Balance without Draft Shield |
12130.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16442555
|
Sartorius
MCA1202S-2S00-0 QP99 |
1200 g | 10 mg | Balance without Draft Shield |
7690.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16442765
|
Sartorius
MCA1203S-2S00-U QP1 |
1200 g | 1 mg | Balance with Draft Shield U and without QP99 Software |
12680.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16453495
|
Sartorius
MCA2202S-2S00-0 QP99 |
2200 g | 10 mg | Balance without Draft Shield | N/A | ||||
16453675
|
Sartorius
MCA2203P-2S00-E QP99 |
1010 g, 2200 g | 1 mg, 10 mg | Balance with Draft Shield E |
12745.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16453705
|
Sartorius
MCA2203P-2S00-R QP99 |
1010 g, 2200 g | 1 mg, 10 mg | Balance with Draft Shield R |
12745.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16456775
|
Sartorius
MCA623P-2S00-U QP99 |
150 g, 300 g, 620 g | 1 mg, 2 mg, 5 mg | Balance with Draft Shield U |
9945.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16465595
|
Sartorius
MCA5202S-2S00-A QP99 |
5200 g | 10 mg | Balance with Draft Shield A |
14130.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16465625
|
Sartorius
MCA5202S-2S00-I QP99 |
5200 g | 10 mg | Balance with Draft Shield I |
15370.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16465655
|
Sartorius
MCA5202S-2S00-U QP99 |
5200 g | 100 mg | Balance with Draft Shield U |
11300.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16465835
|
Sartorius
MCA5203P-2S00-E QP99 |
5200 g | 1 mg | Balance with Draft Shield E |
14980.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16465865
|
Sartorius
MCA5203P-2S00-R QP99 |
5200 g | 1 mg | Balance with Draft Shield R |
14980.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16472775
|
Sartorius
MCA1203S-2S00-U QP99 |
1200 g | 1 mg | Balance with Draft Shield U |
13770.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16477175
|
Sartorius
MCA8202S-2S00-0 QP99 |
8200 g | 10 mg | Balance without Draft Shield |
10730.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Balance with Type Approval Certificate for China | |
Color Touchscreen | |
Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel |
D-sub connector, 9-pin | |
Internal Calibration | |
100/240 V (± 10%), 50/60 Hz, 1.0 A |