Learn More
Sartorius™ Cubis™ II Semi-Micro Lab Balance
BIOPHARMA
Used for weighing small sample amounts down to the μg-range, this balance offers a readability of 10 μg and a maximum weighing capacity of up to 220 g.
Brand:  Sartorius™ MCA225S-2CEU-I
Product Code. 16309655
  • 23760.00 € / Each
Explore more special offers
Description
Description
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Sartorius™ Cubis™ II balance series.
  • All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems
  • Design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement
  • Modern user interfaces and easy process integration
  • Draft Shield I
  • Pharmaceutical and GxP compliance
  • Data handling, integrity and connectivity
  • Ergonomic sample handling
  • Unlimited communication at the highest level of accuracy and precision
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Type Approval Certificate for China, CE
220 g
0.01 mg
2 s
100/240 VAC (±10%), 50/60 Hz, 1 A
D-sub Connector, 9-pin
Color Touchscreen
Aluminum, PBT, Optiwhite Float Glass, Stainless Steel
Analytical Balance
No
Product Suggestions
Product Suggestions
Videos
Videos
  • Sartorius Cubis analytical balances
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ Cubis™ II Semi-Micro Lab Balance > Cubis II Semi-Micro Lab Balance; Capacity: 220 g; Display Type: Color Touchscreen; Readability: 0.01 mg; Stabilization Time: 2 s; Certifications/Compliance: Type Approval Certificate for China, CE