Learn More
Sartorius Cubis™ II Semi-Micro Lab Balance
BIOPHARMA
The Cubis™ II Semi-Micro Lab Balance offers readability of 10 μg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the μg-range.
12200.00 € - 24845.00 €
Specifications
Certifications/Compliance Balance with Type Approval Certificate for China
Display Type Color Touchscreen
Stabilization Time 2 sec.
Type Microbalance
View More Specs
Products 7
Product Code Brand Capacity Readability Includes Price Quantity & Availability  
Product Code Brand Capacity Readability Includes Price Quantity & Availability  
16483075
BIOPHARMA
View Documents
Sartorius
MCA125P-2S00-U QP1
60 g, 120 g 0.01, 0.1 mg Draft Shield U, QP1
12200.00 €
Each
16293182
View Documents
Sartorius
MCA125S-2CEU-I QP99
120 g 0.01 mg Draft Shield I, QP99 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16454315
BIOPHARMA
View Documents
Sartorius
MCA225S-2S00-I QP1
220 g 0.01 mg Draft Shield I, QP1
23210.00 €
Each
16273202
BIOPHARMA
View Documents
Sartorius
MCA225S-2CEU-I QP99
220 g 0.01 mg Draft Shield I, QP99
24845.00 €
1 unit
16293202
BIOPHARMA
View Documents
Sartorius
MCA225S-2CEU-U QP99
220 g 0.01 mg Draft Shield U, QP99
20765.00 €
1 unit
16689881
View Documents
Sartorius
MCA225P-2CEU-I
220 g 0.01, 0.02, 0.05 mg Draft Shield I
16965.00 €
Each
16474195
BIOPHARMA
View Documents
Sartorius
MCA225P-2S00-U QP1
220 g 0.01, 0.02, 0.05 mg Draft Shield U, QP1
14485.00 €
Each
Description
Description
The New Generation of Modular Premium Laboratory Balances
  • Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
  • One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
  • All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
  • These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
  • It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Specifications
Balance with Type Approval Certificate for China
Color Touchscreen
2 sec.
Microbalance