Learn More
R&D Systems™ Cultrex 3D Culture Matrix Rat Collagen I
Applications: Bioactivity
Brand:  R&D Systems™ 3447-020-01
209.06 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18481330
  • 266.00 € / 100mg
Explore more special offers
Description
Description
Functional Assays: Promotes cell attachment and spreading of HT-1080 human fibrosarcoma cells. Collagen I promotes attachment and growth of murine endothelial SVEC4-10 cells in 3-D culture. Gelling Assay: Type I collagen forms a firm gel at neutral pH and 37°C when diluted to 0.4 mg/ml.
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
Specifications
Specifications
Product is stable for a minimum of 3 months if stored at 4 °C. Do Not Freeze. Storage Buffer: 20 mM Acetic Acid Sterility Testing: No bacterial or fungal growth detected after incubation at 37 °C for 14 days following USP XXIV Chapter 71 sterility test. No mycoplasma contamination detected by PCR.
<20.0EU per 1μg of the protein by the LAL method
RUO
Collagen I insoluble extracellular glycoproteins
100 mg
Rat tail tendons
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not For Use In Diagnostic Procedures