missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Poly-L-Lysine (2 x 100 mL)

Applications: Bioactivity

Brand:  R&D Systems™ 3438-200-01

176.24 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18757257

  • 225.00 € / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applications: Substrate for cell culture adhesion. An area of 25 cm2 can be coated with 0.5 ml of a 0.01% Poly-L-Lysine solution. Optimal conditions for attachment must be determined for each cell line and application. Slides may be dipped in the solution and air dried before applying sample. Quality Control: Tested for ability to promote attachment of rat PC-12 pheochromocytoma cells. Cell Culture Qualified: No bacterial or fungal growth detected after incubation at 37degreesC for 14 days following USP XXIV, Chapter 71 sterility testing. No mycoplasma contamination detected by PCR.
Specifications

Specifications

Product is stable for at least 6 months from the date of receipt when stored at 2 - 8° C. Keep sterile.
Poly-L-Lysine
<20.0 EU per 1μg of the protein by the LAL method
Bioactivity
RUO
Liquid
Sterile-filtered
0.01% in Phosphate Buffered Saline, sterile-filtered.
200 mL (2 x 100 mL)
Fermentation using starch.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers