Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Brand:  R&D Systems™ BME001-05
 View more versions of this product
Product Code. 18777896
  • 412.00 € / 5mL
Explore more special offers
Specifications
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
5 mL