Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Brand:  R&D Systems™ BME001-01
 View more versions of this product
Product Code. 18779546
  • 102.00 € / 1mL
Explore more special offers
Specifications
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
1 mL