missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck

Culture your tissues using the Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck. It is designed for consistency, cell health and reproducability with surface and ancillary options you're looking for in a issue culture flask.

Brand:  Fisherbrand™ FB55901

 View more versions of this product

Product Code. 11532283

  • 40.15 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Fluted neck
  • For adherent or suspension cells
  • 1/3 turn cap enables one-handed operation
Specifications

Specifications

250 mL
Fluted
85 mm
Borosilicate Glass
Conical
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck > Capacity: 250mL; Outer Diameter: 85mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.