missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Expression Medium
GREENER_CHOICE

A GMP-manufactured, chemically-defined, serum-free, protein-free, animal origin–free medium specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells.

572.00 € - 9470.00 €

Specifications

Content And Storage Storage conditions: in the dark at 2-8°C
Shipping conditions: ambient
Shelf life: 12 months from date of manufacture
Cell Line ExpiCHO-S™, CHO-S, SFM Adapted
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Suspension Cell Culture
Form Liquid
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15421783
GREENER_CHOICE
View Documents
Gibco™
A2910001
1000 mL
572.00 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
15492163
GREENER_CHOICE
View Documents
Gibco™
A2910002
6 x 1 L
2880.00 €
6 L
Please to purchase this item. Need a web account? Register with us today!
15471044
View Documents
Gibco™
A2910004
20 L
9470.00 €
20 L
Please to purchase this item. Need a web account? Register with us today!
15461044
View Documents
Gibco™
A2910003
10 L
4895.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ExpiCHO Expression Medium is specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells. This medium is GMP-manufactured, chemically-defined, serum-free, protein-free, and animal origin–free. It is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) for scalable transient protein production. To achieve the highest levels of transient protein expression, the medium is matched to a culture feed, ExpiCHO Feed, also included in the system kit and transfection kits.

With ExpiCHO Expression Medium you can:

  • Grow suspension CHO cultures to densities of over 2 x 107 cells per mL, with >95% cell viability
  • Achieve high transfection efficiencies of >90% when coupled with ExpiFectamine CHO transfection reagent
  • Transiently transfect suspension CHO cells at a density of 6 x 106 cells per mL
  • Achieve yields up to 3 g per L of recombinant protein of high-density transiently transfected cultures by sustained, high-level expression
  • Grow and transfect CHO cells at volumes of <1 mL up to 5 L in culture formats from multiwell plates up to 5 L shake flask

ExpiCHO Expression Medium is ready to use. It is formulated with GlutaMAX-I reagent and does not require supplementation. It does not contain phenol red. The chemically-defined, animal origin free formulation results in high reproducibility and lot-to-lot reliability. ExpiCHO Expression Medium is not recommended for adherent cell cultures.

Specifications

Specifications

Storage conditions: in the dark at 2-8°C
Shipping conditions: ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Liquid
Serum-free
No Phenol Red
Room Temperature
ExpiCHO-S™, CHO-S, SFM Adapted
Suspension Cell Culture
CHO Expression Medium
GlutaMAX
ExpiCHO™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.